Fatty acid library plates, SCREEN-WELL®
Supplier: Enzo Life Sciences
ALEXBML-2803-0500EA 2740 EUR
ALEXBML-2803-0500 ALEXBML-2803-0100
Fatty acid library plates, SCREEN-WELL®
Microplates HTS Microplates
SCREEN-WELL® Fatty Acid Library contains 64 fatty acids of varying saturation and chain length.
A diverse set of fatty acids can be a rich source of compounds for receptor de-orphaning, inhibitor screening, and high content screening. This set contains both fatty acids with defined activities and fatty acids with putative biological activities.Library includes: Unsaturated & saturated fatty acids, arachidonic acid, C10-24 chain length linolenic acid, E and Z isomers, γ-linolenic acid, and many more.
Fatty acids are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates. Concentration of 10 mM in DMSO.