N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (by HPLC)

Supplier: Enzo Life Sciences

ALEXBML-EI178-0050EA 138 EUR
ALEXBML-EI178-0050 ALEXBML-EI178-0250
N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (by HPLC)
N-(4-n-Pentylcinnamoyl)anthranilic acid

ACA is a close structural analog of the PLA2 inhibitor, ONO-RS-082 and possesses similar but less potent inhibitory activity. Inhibits TRP channels independently of PLA2, including TRPM2, TRPC6 and TRPM8. It is a useful tool for studying the physiological role of TRP channels.


  • Inhibitor of PLA2 and TRP channels


White solid.


Soluble in DMSO (50 mg/ml) or ethanol (50 mg/ml @ 60°C).

Formula: C₂₁H₂₃NO₃
MW: 337,419 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD00211365
CAS Number: 110683-10-8
https://export.vwr.com/stibo/web/std.lang.all/36/75/45403675.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR