Lycorine HCl ≥97%

Supplier: Enzo Life Sciences

ALEXBML-GR313-0010EA 140 EUR
ALEXBML-GR313-0010 ALEXBML-GR313-0050
Lycorine HCl ≥97%
Lycorine HCl

Lycorine is a eukaryotic termination inhibitor.


  • Protein synthesis inhibitor


White to off-white powder.


Soluble in water (>15mg/ml).

Formula: C₁₆H₁₇NO₄·HCl
MW: 323,8 g/mol
Storage Temperature: Freezer
CAS Number: 2188-68-3
https://export.vwr.com/stibo/web/std.lang.all/39/51/45403951.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR