Anti-Endothelin B Receptor Rabbit Polyclonal Antibody

Supplier: Axxora
ALEXBML-SA577-0050EA 420 EUR
ALEXBML-SA577-0050
Anti-Endothelin B Receptor Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes rat ET-B.

Type: Primary
Antigen: ENDOTHELIN B RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR