Jurkat cell extract (induced)
Supplier: Enzo Life Sciences
ALEXBML-SW116-0100EA 54.6 EUR
ALEXBML-SW116-0100
Jurkat cell extract (induced)
Cells
Whole cell extract of human Jurkat macrophage cells, induced to undergo apoptosis by the chemotherapeutic agent etoposide.
  • Formulation: Liquid, cell extract in SDS-PAGE sample buffer
  • Ready to use
WB control for PARP antibodies to detect intact 116 kDA PARP and 85 kDa apoptosis-related cleavage product. Also used for detection of -procaspase-3 processing to active enzyme using caspase-3 antibody; and pro-caspase-9. Recommended usage is 10 µl per lane.