Vincristine sulphate ≥98%

Supplier: Enzo Life Sciences

ALEXBML-T117-0005EA 199 EUR
ALEXBML-T117-0005 ALEXBML-T117-0025
Vincristine sulphate ≥98%
Vincristine sulphate

Microtubule inhibitor


Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Depolymerizes microtubules and blocks binding of tubulin to microtubule proteins. Causes aberrant tubulin polymerization. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Posttranscriptionally downregulates HIF-1α expression.

Formula: C₄₆H₅₈N₄O₁₄S
MW: 923,04 g/mol
CAS Number: 2068-78-2
https://export.vwr.com/stibo/web/std.lang.all/15/54/45401554.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR