Anti-Exendin-4 Mouse Monoclonal Antibody [clone: 35]

Supplier: Axxora
ALEXBPDABS012351EA 5270 EUR
ALEXBPDABS012351
Anti-Exendin-4 Mouse Monoclonal Antibody [clone: 35]
Antibodies
Exendin-4 is a 39-amino-acid peptide found in venom from the Gila monster Heloderma suspectum. It is a member of the glucagon-secretin family of peptide hormones and neuropeptides. Exendin-4 is a potent agonist of the GLP-1 receptor, and hence a potent stimulator of insulin secretion.

Type: Primary
Antigen: Exendin-4
Clonality: Monoclonal
Clone: 35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Gila monster (Heloderma suspectum)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR