Cyanine 5-UTP (enhanced)
Supplier: Enzo Life Sciences
ALEXENZ-42506EA 577 EUR
ALEXENZ-42506
Cyanine 5-UTP (enhanced)
Nucleic Acid Reagents Nucleotides
Cyanine 5-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP.