DNA isolation kit, EPIXTRACT®
Supplier: Enzo Life Sciences
EpiXtract®
ALEXENZ-45018-0100EA 443 EUR
ALEXENZ-45018-0100 ALEXENZ-45018-0050
DNA isolation kit, EPIXTRACT®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
EPIXTRACT® DNA isolation kit provides a simple method to isolate DNA from plasma, serum, and body fluids.
  • Rapid, provide results in less than 15 minutes
  • Safe to use, contains only non-toxic reagents and no phenol chloroform
  • Highly efficient, isolate DNA from plasma or serum containing as low as 0,1 ng DNA
Delivery information: Supplied with digestion solution, digestion powder, DNA isolation buffer, DNA elution solution, spin column and collection tube.