KU55933 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

ALEXENZCHM1500010EA 180 EUR
ALEXENZCHM1500010 ALEXENZCHM1500005
KU55933 ≥99% (by HPLC)
KU55933

Potent and selective ATP-competitive inhibitor of ATM


KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer.

Formula: C₂₁H₁₇NO₃S₂
MW: 395,49 g/mol
MDL Number: MFCD08276985
CAS Number: 587871-26-9
https://export.vwr.com/stibo/web/std.lang.all/16/73/45401673.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR