Bi-metallic dial thermometers
Supplier: Amarell
AMARLF105652EA 8.7 EUR
AMARLF105652 AMARLF105650
Bi-metallic dial thermometers
Thermometers Analogue Thermometers
These dial thermometers use a bi-metallic tip.
  • Dishwasher-safe
  • Suitable for core temperatures
  • Clear scale together with a robust design
Division: 1,0 °C
Stainless steel piercing probe