Deep well plates, 24-well, Axygen®
Supplier: Corning
Axygen®
AXYGPDW10ML24C2BCSEA 565 EUR
AXYGPDW10ML24C2BCS AXYGP-DW10ML24CS AXYGP-DW10ML24C
Deep well plates, 24-well, Axygen®
Microplates Deep Well Microplates
Deep well and assay plates are used for sample collection, in vitro growth chambers and longterm storage. Made of PP, they offer excellent chemical resistance and will withstand temperatures down to –80 °C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear V-shaped well bottom.