Reagent bottles, wide neck, with screw cap, Corning®
Supplier: Corning
Corning®
CORN1500P-250EA 92.19 EUR
CORN1500P-250 CORN1500P-1L CORN1500P-2L CORN1500P-500
Reagent bottles, wide neck, with screw cap, Corning®
Bottles Plastic Bottles
PP, translucent, with PP screw cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
Can be used anywhere in the laboratory, as well as for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.