Standard polystyrene microplates, 1536-well, Corning®
Supplier: Corning
Corning®
734-4162EA 2002.79 EUR
734-4162 734-4163 CORN7249 CORN7246 738-0149 CORN7248 CORN7247 734-4128 734-4129 734-1670 734-4125 734-4126 734-4127 734-4164 734-1199 734-1198 734-1671 CORN3831 CORN3838BC 734-4130 CORN3835 CORN3833 CORN3832 738-0150 734-4135 CORN3838 CORN3836 734-4131 CORN3832BC 734-4132 734-4133 734-4134
Standard polystyrene microplates, 1536-well, Corning®
Microplates HTS Microplates
These PS microplates conform to standard microplate footprint and dimensions, and are available in solid black or white, with round or flat bottoms, or in black clear bottom formats.
  • Total well volume of 10 μl for round well plates and 12,8 μl for flat bottom plates
  • Recommended working volume of up to 8 μl
  • Round well bottoms for reduced air entrapment and improved CVs and Z factor
  • Raised well bottoms for higher sensitivity
  • Lids available separately
* NBS™ = Non binding surface