ThermalTray™, thermoconductive modules
Supplier: Corning
Corning®
CORN432075EA 451 EUR
CORN432075 CORN432074 CORN432073
ThermalTray™, thermoconductive modules
Racks Cooling Racks
ThermalTray thermo-conductive platforms support CoolRack and CoolSink sample modules in liquid temperature sources - such as melting ice, water baths and liquid nitrogen.
  • The stable, sturdy design makes them ideal for processing temperature-sensitive samples
Made of a highly conductive alloy, ThermalTray conducts the source temperature to the CoolRack or CoolSink, and ultimately to your sample.