Corning® media bottles, PET, with IATA-validated screw cap
Supplier: Corning
Corning®
CORN432334EA 280.46 EUR
CORN432334 CORN432333 CORN432332 CORN432331
Corning® media bottles, PET, with IATA-validated screw cap
Bottles Plastic Bottles
Corning® PET media bottles are equipped with a screw cap designed to ensure leak-proof performance.
  • Available in four sizes
  • Packaged in convenient, shrink-wrapped trays within an outer bag to ensure cleanliness
  • Break-resistant bottles are an ergonomic and easy-to-handle alternative to glass
  • Molded graduations for accurate measurements
  • Innovative HDPE screw cap meets IATA requirements to ensure leak-proof air transport conditions
These bottles with IATA-validated screw caps are designed for safe, secure storage of tissue culture media, sera, buffers, and other aqueous solutions. The PET resin makes the bottle lightweight, break-resistant, and as transparent as glass. The screw cap is designed to ensure leak-proof performance and meets International Air Transport Association (IATA) requirements for primary packaging used in air transportation.
Certifications: Bottles have Sterility Assurance Level (SAL) of 10⁻⁶. Bottles are validated USP Class VI, noncytotoxic, nonhemolytic, and nonpyrogenic.