Corning® 1536 well cyclic olefin copolymer (COC) microplate
Supplier: Corning
Corning®
CORN4565EA 1143.45 EUR
CORN4565 CORN4568 CORN4572 CORN4561 CORN4573 CORN4563 CORN4564 CORN4560 CORN4571
Corning® 1536 well cyclic olefin copolymer (COC) microplate
Microplates Cell Culture Plates
Cyclic olefin copolymer (COC). Superior flatness of the plates minimises variability within a plate and between plates. The barcoded plates come with with raised well bottoms for higher sensitivity.
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Working volume is up to 10 µl
  • Flat bottom wells are compatible with automated dispensers