3D Clear tissue clearing reagents and buffers, Corning®
Supplier: Corning
Corning®
CORN5735EA 204.55 EUR
CORN5735 CORN5736 CORN5737 CORN5738 CORN5731 CORN5732 CORN5733 CORN5734 CORN5740 CORN5741 CORN5730 CORN5739
3D Clear tissue clearing reagents and buffers, Corning®
Cell Culture Supplements and Additives
In our Starter Kit we have combined 30 ml of Corning 3D Clear with the four buffer solutions required to process 3D tissue cultures for immunolabelling. All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models.
  • Rapid tissue clearing
  • Easy to use
  • No special equipment required
  • Compatible with IF, FP and other fluorescent labels
  • Microplate and automation compatible
  • Reversible to follow up with 2D H&E/IHC
The Corning 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high throughput processing. When paired with fluorescent labelling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning 3D Clear reagent allows for complete 3D cell culture model characterisation and more accurate drug screening.