Bottle pourers
Supplier: Cowie
COWI028.045BLUEEA 83.8 EUR
COWI028.045BLUE COWI028.030 COWI028.038 COWI028.032
Bottle pourers
Funnels Dropping Funnels
Manufactured in pure, FDA approved, PTFE.
  • Totally chemically resistant and inert
  • To fit GL45, 38, 32 and 30 screw neck bottles
  • State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.