Jerricans, polyethlene
Supplier: Düperthal
DUEH5.11.010EA 287.98 EUR
DUEH5.11.010 DUEH5.09.010
Jerricans, polyethlene
Bottles Jerricans
PE, electrostatically conductive.
  • Good chemical resistance
  • Good temperature resistance
  • Recyclable