VWR®, Casein

Supplier: VWR
VWR®
E666-500GEA 277 EUR
E666-500G
VWR®, Casein
Proteins and Peptides
High purity grade milk protein powder.

  • Loss on drying: 10%
  • Purity: 95%
  • Residue on ignition: 3%
  • Solubility (2,5%, 0,02 N NaOH): PASS
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR