PCR workstation, CaptairBio
Supplier: Erlab
ERLAE20EA 2890 EUR
ERLAE20
PCR workstation, CaptairBio
Fume Hoods and Enclosures PCR Workstation Enclosures
Designed to perform non-pathogenic handlings, these enclosures has been created for handlings involving a high sample cross contamination risk such as sample preparations before thermocycling, Post PCR DNA sequencing revelation / separation, in vitro fertilization, cellular cultures, vegetal biology, preparation of sterile solutions.
  • The worksurface made of stainless steel makes the cleaning easy
  • Total openings for an easy access into the enclosure
  • Energy efficient
  • Automatic UV cut-off in case of front door opening
  • Internal reflectors: no shadow area
Enclosure design from 800 mm to 1,8 m - equipped with a powerful UV decontamination preventing any handling from cross contamination risks.
Optional work surface in stainless steel 304 l with built-in retention tray or tempered glass work surface with sealed frame and rolling or fixed benches used (these must be ordered separately).