Microbiological safety cabinets, class II, S@feMate EZ
Supplier: EuroClone
SAFEMATE™
EUCLLDK400NEA 6990 EUR
EUCLLDK400N EUCLLDK600N
Microbiological safety cabinets, class II, S@feMate EZ
Fume Hoods and Enclosures Biological Safety Cabinets
The S@feMate range of biological safety cabinets, Class II provide an extremely reliable level of safety for the operator, the environment and the product. Including premium features in an entry level priced cabinet, the new S@feMate EZ series allow everybody to stay safe in an easy way.
  • Air and aerosol tight electrical sliding sash: the front glass is operated using the switches on the main control panel allowing effortless opening and closing of the working area
  • The “V”-shaped antiobstruction grill allows working without worrying about armrests, while the self positioning electrically operated front sash makes sure that the front aperture is always at the right size for comfort and safety
  • Real time visualization of airflows speeds
  • State of the art AC motorblower enhances energy efficiency, reducing operating costs
  • 5° Sloping front aperture to maximise comfort
  • UV lamp on back wall (standard)
  • Optional utilities for gas and vacuum, installable in-field
The main structure is made from cold rolled steel, stove enamel coated RAL 7035 + ABS plastics.
The working surface is made from stainless steel AISI 304 - 2B finishing.
Front widow is in laminated safety glass.
Certifications: Protection guaranteed as required by EN12469:2000 standard.
Delivery information: Standard utilities included with the cabinets: 2 Schuko/EuroPlug socket, voltage free contact, DOP test port, UV lamp backwall mounted.
Delivery prices and installation on request.