Microbiological safety cabinets, SAFEMATE CYTO
Supplier: EuroClone
135-1431EA 0 EUR
135-1431 EUCLLY40000
Microbiological safety cabinets, SAFEMATE CYTO
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
These microbiological safety cabinets offer maximum safety for the handling of toxic and dangerous compounds such as cytostatics, isotopes etc.
  • Latest generation microprocessor control
  • Large, high resolution, digital screen
  • Hermetically sealed electric front window with automatic opening in working position
  • Alarm for insufficient air level and incorrect positioning of the front window
  • Exclusive 'Bag-in Bag-out' system for replacement of the tertiary filter (eliminates the risk of the plenum and work cabinet becoming contaminated when the filter is replaced)
Certifications: Compliant with DIN 12980:2005 and EN 12469:2000 standards
Two year warranty extendable to five years with maintenance contract
Delivery information: Please contact your local Avantor sales office for details of delivery and commissioning costs.