Petri dishes, Gosselin™
Supplier: Corning Life Sciences
Gosselin
391-0569EA 306 EUR
391-0569 391-0568 GOSSBP140-02 GOSSBP93B-101 GOSSSB93-102 391-0571 391-0567 GOSSBP143-07 391-2068 GOSSBP90B-102 GOSSBP93D-102 GOSSBP143-06 391-0566 391-0572 391-0570
Petri dishes, Gosselin™
Culture Dishes Petri Dishes
Transparent PS Petri dishes for food industry laboratories.
  • Excellent optical quality
  • Stable stacking due to the stacking ring
  • High level of mechanical resistance, will retain shape up to +55 °C
These dishes are produced in a Class 100 (ISO 5) manufacturing environment, ensuring an aseptic product. Product is labelled with a batch number on each carton. Also available sterile (Sterility Assurance Level based on ISO 11137 : SAL 10-6).