Bottles, narrow neck, square, HDPE, with screw caps
Supplier: Gosselin
GOSSFC1000-50EA 153 EUR
GOSSFC1000-50 GOSSFC1000-51 GOSSFC1000-52
Bottles, narrow neck, square, HDPE, with screw caps
Bottles Plastic Bottles
Made of HDPE, translucent, with PP screw cap.
  • Graduated
  • Good chemical and temperature resistance
  • Tamper evident screw cap with shaped seal
  • Screw cap assembled