Bottles, narrow neck, square, HDPE, with hinged cap
Supplier: Gosselin
GOSSFC510-03EA 152 EUR
GOSSFC510-03 GOSSFC510-02 GOSSFC510-01
Bottles, narrow neck, square, HDPE, with hinged cap
Bottles Plastic Bottles
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance