Bottles, wide neck, square, with screw cap, 100 to 500 ml capacity
Supplier: Gosselin
GOSSFCP100-05EA 442 EUR
GOSSFCP100-05 GOSSLC500-05 GOSSLC250-06 GOSSLC250-03 GOSSLC500-03 GOSSLC500-04 GOSSFCP100-04 GOSSLC500-23 215-0717 215-0714 GOSSFC520-10 215-0735 215-0716 GOSSFC520-04 GOSSFC520-02 GOSSFC520-08 GOSSFC520-06 215-3258 215-3257 215-3256 215-3253 215-3274 215-1293 GOSSLC150A-05 215-3252 215-3273 GOSSLC150A-03 215-3272 GOSSLC520-02 GOSSLC500-16 GOSSLC500-17 GOSSFCP500-03 GOSSLC500-10 GOSSFC500-12 GOSSLC500-11 215-0732 215-0734 215-3249 215-0730 215-3961 215-3960
Bottles, wide neck, square, with screw cap, 100 to 500 ml capacity
Bottles Plastic Bottles
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
  • Good chemical resistance, good temperature resistance
  • Sterile oder packed under aseptic conditions
  • Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.