Containers, wide mouth, with hinged cap, Corning® Gosselin™
Supplier: Corning
Corning®
215-3270EA 278 EUR
215-3270 GOSSFL100-05 GOSSFL60-04 215-3269 GOSSFL300-04 215-1285
Containers, wide mouth, with hinged cap, Corning® Gosselin™
Sample Containers General Purpose Containers
Made of PP, natural or coloured.
  • Available in sterile or under aseptic conditions packed versions
  • Very good chemical and temperature resistance
  • Graduated
  • Without cap
Ideal for sampling, storage and transport of liquid and solid samples.
Sized to provide adequate volume for milk sampling. It is shock resistant and is assured sterile 10-3.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904