Containers, wide mouth, with hinged cap
Supplier: Gosselin
215-1285EA 145 EUR
215-1285 215-3270 GOSSFL60-04 215-3269 GOSSFL300-04 GOSSFL100-05
Containers, wide mouth, with hinged cap
Sample Containers General Purpose Containers
Made of PP, natural or coloured.
  • Available in sterile or under aseptic conditions packed versions
  • Very good chemical and temperature resistance
  • Graduated
  • Without cap
Ideal for sampling, storage and transport of liquid and solid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904