Jerricans, square or rectangular, HDPE, Corning® Gosselin™
Supplier: Corning
Corning®
GOSSJCP5-03EA 120 EUR
GOSSJCP5-03 GOSSJCP5-01 GOSSJCP2-01 GOSSJCP5-02 GOSSJCP2-02 216-4272 GOSSHJC5-01 GOSSJCP10-02
Jerricans, square or rectangular, HDPE, Corning® Gosselin™
Bottles Jerricans
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.
  • Good chemical resistance
  • Temperature resistant from -40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.