Bottles, water sampling, octogonal
Supplier: Gosselin
GOSSHP250B-53EA 386 EUR
GOSSHP250B-53 GOSSHP500B-50 GOSSP1000B-52 GOSSHP250B-50 GOSSHP500B-51 GOSSP250B-51 GOSSHP500B-52 GOSSP1000B-51 GOSSP500B-51 GOSSHP250B-51 GOSSHP1000B-53 GOSSHP1000B-50
Bottles, water sampling, octogonal
Bottles Water Sampling Bottles
The water samling bottles, made of PET, transparent, with screw cap are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
  • Molded graduations
  • Unbreakable
  • Tamper-evident screw cap