Jerricans, square or rectangular, with screw cap
Supplier: Gosselin
GOSSJCP5-02EA 78.4 EUR
GOSSJCP5-02 GOSSJCP5-03 GOSSJCP5-01 216-4272 GOSSJCP2-02 GOSSJCP10-02 GOSSJCP2-01 GOSSHJC5-01
Jerricans, square or rectangular, with screw cap
Bottles Jerricans
HDPE, translucent
  • Good chemical resistance
  • Temperature resistant from -40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.
  • Good chemical resistance
  • Temperature resistant from -40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.