Storage bottles, octogonal, PET
Supplier: Gosselin
GOSSP125B-50EA 583 EUR
GOSSP125B-50 GOSSP500B-50 GOSSP1000B-06 GOSSP250B-50 GOSSP1000B-50 GOSSP125B-08 GOSSP125B-06
Storage bottles, octogonal, PET
Bottles Plastic Bottles
These storage bottles are made from break resistant polyethylene terephthalate (PET) with transparency equivalent to glass.
  • Molded graduations
  • Ergonomic shape for easy handling
  • Leak proof, tamper evident HDPE screw cap
Available with Sterility Assurance Level (SAL) of 10⁻³.