Bottles, water sampling, octogonal, Corning® Gosselin™
Supplier: Corning
Corning®
GOSSHP250B-53EA 447 EUR
GOSSHP250B-53 GOSSHP250B-51 GOSSHP1000B-53 GOSSHP250B-50 GOSSHP500B-50 GOSSHP500B-51 GOSSP250B-51 GOSSP1000B-51 GOSSP500B-51
Bottles, water sampling, octogonal, Corning® Gosselin™
Bottles Water Sampling Bottles
The water samling bottles, made of PET, transparent, with screw cap are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
  • Molded graduations
  • Unbreakable
  • Tamper-evident screw cap