Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Corning®
GOSSTP53C-014EA 199 EUR
GOSSTP53C-014 GOSSTP53C-032 GOSSTP53C-052 GOSSTP53C-030 216-2090 216-2094 216-2093 216-2092 216-2091 216-2098 216-2097 216-2096 216-2095 216-2113 GOSSTP30C-003 GOSSTP30C-004 216-2111 216-2099 GOSSTP30C-021 216-0518 GOSSTP53C-023 GOSSTP175-06 GOSSTP53C-021 GOSSTP53C-042 216-2087 216-2086 216-2085 216-2084 216-2102 GOSSTP30C-014 215-1324 216-2100 216-2089 216-2088 GOSSTP30C-032 216-2104 216-2107
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Sample Containers General Purpose Containers
PP translucent or PS clear, with PE screw cap. Straight walled vessels for sample collection, storage and transport. Twist closure for quick opening and closing, with leakproof inner lip. PS offers good thermal and chemical resistance. PP offers excellent thermal and chemical resistance.
  • Selection of colour coded closures for easier sample identification
  • Available sterile or packed under aseptic conditions
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904