Sample dippers
Supplier: Greiner bio-one
GREI224140_50EA 94.3 EUR
GREI224140_50
Sample dippers
Samplers Sample Dippers
Sample dippers designed for safe sampling in food manufacturing.
  • Manufactured from chemically inert PS
Suitable for pre-analytical applications.
Packaging: Individually packed.