Strips of 8 PCR tubes without cap
Supplier: Greiner bio-one
GREI673277_125EA 134 EUR
GREI673277_125 GREI673210_125 GREI673210_1250 GREI373275_125 GREI673275_125 GREI673273_125 GREI373277_125 GREI373270_1250 GREI373273_125 GREI673271_1250 GREI373250_1250 GREI673276_125 GREI373274_125 GREI373276_125 GREI673274_125
Strips of 8 PCR tubes without cap
Tubes PCR Tubes
PP, translucent, coloured.
  • Excellent temperature resistance
  • Excellent chemical resistance
  • Non sterile
  • For single use only
Sappphire PCR 8-tube strips, 0,2 ml, without cap