Conductive pipette tips
Supplier: Greiner bio-one
GREI686296_5000EA 0 EUR
GREI686296_5000 GREI686288_600 GREI764280_7200
Conductive pipette tips
Pipette Tips
Conductive tips are used to prevent carryover in automated pipetting robots. The addition of graphite to the raw material PP makes the pipette tips electrically conductive. By measuring electric currents, the filling level can be determined in sample and reagent containers, so that the depth of immersion of the tip can be adjusted to the filling level.
  • Filter tip: Free of detectable DNase, RNase, human DNA; non pyrogenic, non cytotoxic