Gel loading pipette tips
Supplier: Greiner bio-one
GREI770280_960EA 0 EUR
GREI770280_960 GREI770291_3840
Gel loading pipette tips
Pipette Tips
These gel loading tips are characterised by an elongated end. They are ideal for numerous molecular biological applications, especially for filling the slots of sequencing, PCR and protein electrophoresis gels.
  • Free of detectable DNase, RNase and human DNA
  • Non pyrogenic
  • Non cytotoxic