Pipette tips, Sapphire
Supplier: Greiner bio-one
GREI771255_960EA 94.3 EUR
GREI771255_960 GREI737257_960 GREI771258_960
Pipette tips, Sapphire
Pipette Tips
These tips are manufactured from virgin materials. All tips are transparent, graduated, and allow precise and comfortable pipetting with maximal recovery.
  • Graduations for perfect visual control of liquid
  • Thin-walled top for optimal seal
  • Free of detectable DNase, RNase, human DNA and non pyrogenic
  • Available as low retention tips for even higher recovery