Containers for plant tissue culture
Supplier: Greiner bio-one
216-3451EA 380 EUR
216-3451 212-3219 GREI968162_168 GREI960161_300 GREI968177_192
Containers for plant tissue culture
Tubes Culture Tubes
Container, PS, clear for plant tissue culture. These containers are specially suited for plant tissue culture. The use of a very clear material ensures maximum light transmission and thus rapid and successful growth. These culture containers are not only suitable for the proliferation of plant tissue cultures but can also be used as transport containers.
  • Available with or without lid
  • Polystyrene for high clarity
  • ith flat bottom
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904