Medical mask, IIR
Supplier: Xiantao Yongli
113-9929EA 8.1 EUR
113-9929 113-9924
Medical mask, IIR
Masks Surgical Masks
These medical face masks are made from a soft and flexible non woven fabric, and are free of natural latex.
  • Excellent filter performance >99% BFE
  • 3-ply design
  • Latex-free elastic bands
  • Nose bridge
  • Guaranteed to meet 4.0 AQL level II
The 3-ply material construction features an 25 gsm SPP exterior, 25 gsm MB middle layer, and 25 gsm SPP interior. Colour: Blue.
L×W (unfolded): 170×90 mm
Certifications: Complies with EN 14683:2019 Type IIR requirements.