Nitrile gloves, white, PUREZERO® HG3
Supplier: Halyard
Purezero®
112-6530EA 506 EUR
112-6530 112-6531 112-6510 112-6511 112-6512 112-6513 112-6524 112-6525 112-6526 112-6527 112-6528 112-6529 112-6509
Nitrile gloves, white, PUREZERO® HG3
Gloves
PUREZERO® nitrile cleanroom gloves are made by an accelerator-free formulation which reduces the risk of allergies and skin irritation associated with accelerator chemicals in other nitrile gloves. As a result, PUREZERO® gloves are comfortable to wear, allowing workers to focus on their delicate tasks rather than their gloves.
  • Designed to protect workers with effective barrier protection against chemical splash, microorganisms and viruses
  • Clean-processed to ensure consistent control of low particles and extractables
  • Recommended for use in ISO Class 3 cleanrooms or higher
  • Tacky outer glove finish
  • AQL is 1,5
Gloves are not formulated with these commonly used vulcanising chemicals: Sulphur, thiurams, thiaxoles, guanidines and carbamates.
Certifications: EN ISO 374-1:2016/Type C (K), EN ISO 374-5:2016 Microorganism and Virus Protection, EN ISO 374-4; Cat. III