Scalpel blades
Supplier: Karl Hammacher GmbH
HAMMHSB931-08EA 103 EUR
HAMMHSB931-08 HAMMHSB925-05 HAMMHSB921-03 HAMMHSB935-12 HAMMHSB913-20
Scalpel blades
Scalpels and Scalpel Assemblies Scalpel Blades
Non sterile scalpel blades made from stainless steel. For use with handle HSB 910-14.