Scalpels
Supplier: Karl Hammacher GmbH
HAMMHWB223-30EA 65.8 EUR
HAMMHWB223-30 233-0092 233-0093 233-0094 233-0095 HAMMHWB225-40
Scalpels
Scalpels and Scalpel Assemblies Scalpels
These scalpels are made from Wironit steel (18/12 stainless steel with 18% chrome and from 10 to 12% nickel).
  • Resistant to corrosion and anti-magnetic
  • With fixed blade and metal handle