Avantor® Allsep Anion, Anion Exchange HPLC Columns
Supplier: VWR
Avantor® Allsep
HICH51207EA 2020 EUR
HICH51207 38108/P HICH38109 HICH51212 HICH51200 HICH51214 HICH51213 HICH51209 HICH51208 38108/S 38109/S
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Chromatography Columns
Allsep anion is a methacrylate based phase with quaternary ammonium functional groups, optimised for use with both suppressed and non-suppressed conductivity detection. They are recommended for applications involving inorganic anions, weak and strong acid ions, metal complexes and organic acids.
  • pH range of 2 to 10
  • Can be used with 0 to 100% organic modifier
These columns are compatible with common ion chromatography mobile phases, such as carbonate, bicarbonate, p-hydroxybenzoic acid, phthalic acid, succinic acid and sodium octane sulphonate.
Certifications: Meets the requirements for the EPA method 300.0 Part A for determining inorganic ions in water.
Accessories information: Use guard cartridges with holder (80101/N) and coupler (HI-081).