Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Supplier: HyClone products (Cytiva)
HYCLSH30588.02EA 30.5 EUR
HYCLSH30588.02 HYCLSH30268.LS HYCLSH30588.01 HYCLSH30268.02 HYCLSH30030.03 HYCLSH30268.01 HYCLSH30031.FS HYCLSH30015.03 HYCLSH30015.02 HYCLSH30031.01 HYCLSH30030.02 HYCLSH30031.03 HYCLSH30031.02 HYCLSH30016.02
Cell culture buffers, Hank's balanced salt solutions (HBSS) HyClone™
Buffers Cell Culture Buffers
Balanced salt solutions for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Powder formats rehydrate to specified quantity
  • Solutions: 0,1 µm sterile filtered
Buffer cell culture growth with HyClone Hank's Balanced Salt Solutions (HBSS), available in lot sizes that facilitate scale-up and manufacturing.