HyClone™ SFM4MegaVir Cell Culture Media
Supplier: HyClone products (Cytiva)
HYCLSH30587.01EA 261.17 EUR
HYCLSH30587.01
HyClone™ SFM4MegaVir Cell Culture Media
Cell Culture Media
HyClone™ SFM4MegaVir cell culture media is a protein-free medium designed to increase process yields in the manufacture of viral vaccines in a variety of cell lines including Vero, MDCK, COS-7 and MDBK. Manufactured according to cGMP guidelines.
  • Defined protein-free formulation
  • Animal derived component free
  • Developed through Metabolic Pathway Design process for high viral vaccine production
  • Suitable for direct adaptation
  • Designed for a variety of culture applications, including microcarrier cultures