HyperSep™ SpinTip Microscale Solid Phase Extraction Tips
Supplier: Thermo Scientific
HyperSep™
HYPE60109-420EA 0 EUR
HYPE60109-420 HYPE60109-419 HYPE60109-407 HYPE60109-409 HYPE60109-422 HYPE60109-411 HYPE60109-410 HYPE60109-421 HYPE60109-402 HYPE60109-404 HYPE60109-425 HYPE60109-414 HYPE60109-403 HYPE60109-406 HYPE60109-405
HyperSep™ SpinTip Microscale Solid Phase Extraction Tips
Pipette Tips
HyperSep™ SpinTips are a revolutionary, patented series of micropipette tips designed for sample preparation.
  • Eliminates the need for a filter and, thus, dead volume
  • HyperSep™ SpinTip is a unique concept in solid phase extraction (SPE)
  • excellent when working with small samples
These tips feature a fine slit at the bottom (slit width 1 to 2 μm) which permits liquid to pass through, but retains the chromatographic material (20 to 30 μm) in the tip. HyperSep™ SpinTips contain just the desired chromatography material and nothing else.
They are ideal when working with small samples and are available with a variety of materials to suit a wide range of applications.